Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079316.1 | NP_001072784.1 | 3 - 4 | 296 | 71 | |
XM_012954043.2 | XP_012809497.1 | 2 - 3 | 484 | 71 | ||
GenBank mRNA | BC118860.1 | - | 3 - 4 | 296 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |