Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079429.1 | NP_001072897.1 | 6 - 7 | 845 | 74 | |
XM_018092186.1 | XP_017947675.1 | 6 - 7 | 737 | 74 | ||
GenBank mRNA | BC118853.1 | - | 6 - 7 | 845 | 74 | |
BC135261.1 | - | 6 - 7 | 836 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |