Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097177.1 | NP_001090646.1 | 2 - 3 | 278 | 111 | |
XM_012960358.2 | XP_012815812.1 | 3 - 4 | 1261 | 111 | ||
XM_012960360.2 | XP_012815814.1 | 2 - 3 | 1171 | 111 | ||
GenBank mRNA | BC127274.1 | - | 2 - 3 | 278 | 111 | |
CU024990.1 | - | 2 - 3 | 265 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |