Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100214.1 | NP_001093684.1 | 3 - 4 | 423 | 68 | |
GenBank mRNA | BC136136.1 | - | 3 - 4 | 423 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |