Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c18orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102910.1 | NP_001096380.1 | 2 - 3 | 178 | 95 | |
GenBank mRNA | BC135947.1 | - | 2 - 3 | 178 | 95 | |
CR762189.1 | - | 2 - 3 | 255 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |