Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102949.1 | NP_001096419.1 | 2 - 3 | 2403 | 91 | |
GenBank mRNA | BC136083.1 | - | 2 - 3 | 2403 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |