Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105284.1 | NP_001098754.1 | 5 - 6 | 459 | 106 | |
GenBank mRNA | BC152057.1 | - | 5 - 6 | 459 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |