Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112955.1 | NP_001106426.1 | 4 - 5 | 1166 | 104 | |
GenBank mRNA | BC153370.1 | - | 4 - 5 | 1166 | 104 | |
CT027853.1 | - | 4 - 5 | 1233 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |