Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112963.1 | NP_001106434.1 | 9 - 10 | 1301 | 89 | |
GenBank mRNA | BC153720.1 | - | 9 - 10 | 1301 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |