Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112979.1 | NP_001106450.1 | 9 - 10 | 896 | 83 | |
XM_012969782.2 | XP_012825236.1 | 8 - 9 | 867 | 83 | ||
GenBank mRNA | BC153743.1 | - | 9 - 10 | 896 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |