Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113063.1 | NP_001106534.1 | 2 - 3 | 173 | 75 | |
GenBank mRNA | BC154903.1 | - | 2 - 3 | 173 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |