Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113125.1 | NP_001106596.1 | 5 - 6 | 898 | 106 | |
XM_012956373.2 | XP_012811827.1 | 5 - 6 | 963 | 106 | ||
GenBank mRNA | BC155426.1 | - | 5 - 6 | 898 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |