Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113838.1 | NP_001107310.1 | 5 - 6 | 2319 | 67 | |
XM_012962670.2 | XP_012818124.1 | 5 - 6 | 2219 | 67 | ||
XM_012962671.2 | XP_012818125.1 | 5 - 6 | 2179 | 67 | ||
XM_012962672.2 | XP_012818126.1 | 5 - 6 | 2216 | 67 | ||
XM_012962673.2 | XP_012818127.1 | 5 - 6 | 2219 | 67 | ||
XM_012962675.2 | XP_012818129.1 | 5 - 6 | 2216 | 67 | ||
XM_012962676.2 | XP_012818130.1 | 5 - 6 | 2219 | 67 | ||
XM_012962677.2 | XP_012818131.1 | 5 - 6 | 2219 | 67 | ||
GenBank mRNA | BC155706.1 | - | 5 - 6 | 2319 | 67 | |
CT025421.2 | - | 5 - 6 | 2218 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |