Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114031.1 | NP_001107503.1 | 2 - 3 | 195 | 89 | |
XM_012964257.2 | XP_012819711.1 | 246 | 89 | |||
GenBank mRNA | BC157661.1 | - | 2 - 3 | 195 | 89 | |
CR926350.2 | - | 154 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |