Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpini1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123493.1 | NP_001116965.1 | 4 - 5 | 724 | 74 | |
XM_018093779.1 | XP_017949268.1 | 4 - 5 | 770 | 74 | ||
GenBank mRNA | BC160993.1 | - | 4 - 5 | 724 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |