Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126728.1 | NP_001120200.1 | 7 - 8 | 913 | 98 | |
GenBank mRNA | BC159323.1 | - | 7 - 8 | 913 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |