Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cxorf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127142.1 | NP_001120614.1 | 5 - 6 | 1109 | 79 | |
GenBank mRNA | BC161752.1 | - | 5 - 6 | 1109 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |