Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126517.1 | NP_001119989.1 | 4 - 5 | 632 | 68 | |
XM_012956422.2 | XP_012811876.1 | 4 - 5 | 443 | 68 | ||
XM_012956423.2 | XP_012811877.1 | 4 - 5 | 450 | 68 | ||
XM_012956424.2 | XP_012811878.1 | 4 - 5 | 423 | 68 | ||
XM_012956425.2 | XP_012811879.1 | 4 - 5 | 379 | 68 | ||
XM_012956426.1 | XP_012811880.1 | 3 - 4 | 360 | 68 | ||
XM_018091076.1 | XP_017946565.1 | 4 - 5 | 378 | 68 | ||
XM_018091077.1 | XP_017946566.1 | 4 - 5 | 377 | 68 | ||
XM_018091078.1 | XP_017946567.1 | 4 - 5 | 670 | 68 | ||
GenBank mRNA | BC158248.1 | - | 4 - 5 | 632 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |