Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127973.2 | NP_001121445.2 | 4 - 5 | 496 | 68 | |
GenBank mRNA | BC166198.1 | - | 485 | 68 | ||
CX939571.1 | - | 4 - 5 | 496 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |