Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130210.1 | NP_001123682.1 | 2 - 3 | 105 | 95 | |
XM_012953918.1 | XP_012809372.1 | 2 - 3 | 157 | 95 | ||
GenBank mRNA | BC167140.1 | - | 2 - 3 | 105 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |