Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130264.1 | NP_001123736.1 | 8 - 9 | 1333 | 91 | |
GenBank mRNA | BC167387.1 | - | 8 - 9 | 1333 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |