Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130343.3 | NP_001123815.2 | 2 - 3 | 2662 | 73 | |
GenBank mRNA | DT443629.1 | - | 2 - 3 | 313 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |