Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142241.1 | NP_001135713.1 | 5 - 6 | 888 | 86 | |
XM_012956584.2 | XP_012812038.1 | 5 - 6 | 975 | 86 | ||
GenBank mRNA | BC169148.1 | - | 5 - 6 | 888 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |