Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016286.2 | NP_001016286.1 | 3 | 1251 | 103 | |
GenBank mRNA | BC171059.1 | - | 3 | 1224 | 103 | |
CR761595.2 | - | 3 | 1251 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |