Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gja7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205831.1 | NP_991400.1 | 1 | 1051 | 83 | |
GenBank mRNA | BC066131.1 | - | 1 | 1051 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |