Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cesa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112202.1 | NP_001105672.1 | 2681 | 65 | ||
GenBank mRNA | AY372244.1 | - | 2681 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Zea mays |