Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136824.1 | NP_001130296.1 | 669 | 61 | ||
GenBank mRNA | BT033466.1 | - | 669 | 61 | ||
BT033823.1 | - | 695 | 61 | |||
BT054961.1 | - | 669 | 61 | |||
BT066424.1 | - | 669 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |