Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100280820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153740.1 | NP_001147212.1 | 1249 | 79 | ||
GenBank mRNA | BT060611.1 | - | 1238 | 79 | ||
EU953989.1 | - | 1249 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Recommended Assay Zea mays |