Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112092.1 | NP_001105562.1 | 372 | 77 | ||
GenBank mRNA | BT065528.1 | - | 372 | 77 | ||
BT065599.1 | - | 405 | 77 | |||
EU959120.1 | - | 380 | 77 | |||
X55967.1 | - | 372 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |