Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112174.1 | NP_001105644.1 | 124 | 70 | ||
GenBank mRNA | AF366295.1 | - | 124 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |