Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112426.1 | NP_001105896.1 | 726 | 101 | ||
GenBank mRNA | X55981.1 | - | 726 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Zea mays |