Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111928.1 | NP_001105398.1 | 1623 | 68 | ||
GenBank mRNA | HQ858748.1 | - | 1586 | 68 | ||
U81960.1 | - | 1623 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Recommended Assay Zea mays |