Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTCTGACAGTACAAGACATTCA[A/G]TTCTCTTCAAGTCATGTAATCGACT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604371 | |||||||||||||||||||||||
Literature Links: |
HHLA2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HHLA2 - HERV-H LTR-associating 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282556.1 | 2 | Intron | NP_001269485.1 | |||
NM_001282557.1 | 2 | Intron | NP_001269486.1 | |||
NM_001282558.1 | 2 | Intron | NP_001269487.1 | |||
NM_001282559.1 | 2 | Intron | NP_001269488.1 | |||
NM_007072.3 | 2 | UTR 5 | NP_009003.1 | |||
XM_005247080.3 | 2 | Intron | XP_005247137.1 | |||
XM_011512362.1 | 2 | Intron | XP_011510664.1 | |||
XM_011512363.1 | 2 | Intron | XP_011510665.1 | |||
XM_011512364.1 | 2 | Intron | XP_011510666.1 | |||
XM_011512367.2 | 2 | Intron | XP_011510669.1 | |||
XM_017005642.1 | 2 | Intron | XP_016861131.1 |