Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTCTCCCCATCACCATTCGTC[A/G]GGCCTCTGTTTCGTGGTATCATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP8A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP8A1 - ATPase phospholipid transporting 8A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105529.1 | 3670 | Silent Mutation | NP_001098999.1 | |||
NM_006095.2 | 3670 | Silent Mutation | NP_006086.1 | |||
XM_005248043.2 | 3670 | Silent Mutation | XP_005248100.1 | |||
XM_011513615.1 | 3670 | Silent Mutation | XP_011511917.1 | |||
XM_011513616.2 | 3670 | Silent Mutation | XP_011511918.1 | |||
XM_011513618.1 | 3670 | Silent Mutation | XP_011511920.1 | |||
XM_017007645.1 | 3670 | Silent Mutation | XP_016863134.1 | |||
XM_017007646.1 | 3670 | Silent Mutation | XP_016863135.1 | |||
XM_017007647.1 | 3670 | Silent Mutation | XP_016863136.1 |