Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATTTTATACTTAAGGCTGTAAA[C/T]GGCAAAGTCCCATAGATATTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614451 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELOVL7 - ELOVL fatty acid elongase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104558.1 | 4092 | UTR 3 | NP_001098028.1 | |||
NM_001297617.1 | 4092 | UTR 3 | NP_001284546.1 | |||
NM_001297618.1 | 4092 | UTR 3 | NP_001284547.1 | |||
NM_024930.2 | 4092 | UTR 3 | NP_079206.2 | |||
XM_005248606.4 | 4092 | UTR 3 | XP_005248663.1 | |||
XM_005248607.3 | 4092 | UTR 3 | XP_005248664.1 | |||
XM_006714695.3 | 4092 | UTR 3 | XP_006714758.1 | |||
XM_011543651.2 | 4092 | UTR 3 | XP_011541953.1 | |||
XM_017009885.1 | 4092 | UTR 3 | XP_016865374.1 | |||
XM_017009886.1 | 4092 | UTR 3 | XP_016865375.1 | |||
XM_017009887.1 | 4092 | UTR 3 | XP_016865376.1 | |||
XM_017009888.1 | 4092 | Intron | XP_016865377.1 | |||
XM_017009889.1 | 4092 | Intron | XP_016865378.1 |