Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTCAACTAGGTAATTTTAGAA[G/T]TAGATACTTTAAAAAAGAACTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYSMD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYSMD3 - LysM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286812.1 | 4311 | UTR 3 | NP_001273741.1 | |||
NM_198273.2 | 4311 | UTR 3 | NP_938014.1 | |||
XM_005248421.4 | 4311 | UTR 3 | XP_005248478.1 |
POLR3G - RNA polymerase III subunit G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |