Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGTGGGCCTGTAAGGGGAAGAA[C/T]CCAAGAGAACCATTTCCAAGTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERVFRD-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERVFRD-1 - endogenous retrovirus group FRD member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207582.2 | 1929 | Missense Mutation | ATT,GTT | I,V 483 | NP_997465.1 |
SMIM13 - small integral membrane protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135575.1 | 1929 | Intron | NP_001129047.1 |