Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGTCAAAAATCTTGACACGCC[C/T]CCTGCCGTCGTGCACCACCACGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608072 MIM: 187680 | ||||||||||||||||||||
Literature Links: |
NHLRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NHLRC1 - NHL repeat containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198586.2 | 459 | Missense Mutation | AGG,GGG | R,G 149 | NP_940988.2 |
TPMT - thiopurine S-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |