Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACAACCTTTTTTATAAGGCTAA[A/G]CACCAGGAAATCTATAAAATAGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927730 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927730 - uncharacterized LOC101927730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINB9 - serpin family B member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004155.5 | 1804 | UTR 3 | NP_004146.1 | |||
XM_005249184.4 | 1804 | UTR 3 | XP_005249241.1 | |||
XM_011514678.2 | 1804 | UTR 3 | XP_011512980.1 | |||
XM_017010942.1 | 1804 | UTR 3 | XP_016866431.1 | |||
XM_017010943.1 | 1804 | UTR 3 | XP_016866432.1 |