Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTCATGGTGTCTTCATTATTA[C/T]GAAAATCACAGTAATATGACTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENPP5 - ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290072.1 | 2352 | UTR 3 | NP_001277001.1 | |||
NM_001290073.1 | 2352 | UTR 3 | NP_001277002.1 | |||
NM_021572.5 | 2352 | UTR 3 | NP_067547.1 | |||
XM_005249260.3 | 2352 | UTR 3 | XP_005249317.1 | |||
XM_011514786.2 | 2352 | UTR 3 | XP_011513088.1 |