Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATAGGATCTAGTCTAGAGTTGT[A/G]TTCCAGTTGGTTGTTATGTTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608497 MIM: 616251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR2F1 - olfactory receptor family 2 subfamily F member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012369.2 | Intron | NP_036501.2 | ||||
XM_011516019.2 | Intron | XP_011514321.1 | ||||
XM_011516020.2 | Intron | XP_011514322.1 |
TCAF1 - TRPM8 channel associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |