Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCTGAGCTCAGCTGTAAACTT[A/C]GGAGAAGACAGAAAGATCTCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM180A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM180A - family with sequence similarity 180 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205855.3 | 2804 | Intron | NP_995327.1 | |||
XM_006715983.3 | 2804 | UTR 3 | XP_006716046.1 |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |