Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATAACTTGGTTCTTTTTTTTTT[C/T]CTAATCTTGAGATTTCCATAGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX31 - sorting nexin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152628.3 | 1741 | UTR 3 | NP_689841.3 | |||
XM_005250815.3 | 1741 | UTR 3 | XP_005250872.1 | |||
XM_011516899.1 | 1741 | UTR 3 | XP_011515201.1 | |||
XM_017013153.1 | 1741 | UTR 3 | XP_016868642.1 | |||
XM_017013154.1 | 1741 | UTR 3 | XP_016868643.1 | |||
XM_017013155.1 | 1741 | UTR 3 | XP_016868644.1 | |||
XM_017013156.1 | 1741 | UTR 3 | XP_016868645.1 | |||
XM_017013157.1 | 1741 | UTR 3 | XP_016868646.1 | |||
XM_017013158.1 | 1741 | UTR 3 | XP_016868647.1 | |||
XM_017013159.1 | 1741 | UTR 3 | XP_016868648.1 | |||
XM_017013160.1 | 1741 | UTR 3 | XP_016868649.1 | |||
XM_017013161.1 | 1741 | UTR 3 | XP_016868650.1 | |||
XM_017013162.1 | 1741 | UTR 3 | XP_016868651.1 | |||
XM_017013163.1 | 1741 | UTR 3 | XP_016868652.1 | |||
XM_017013164.1 | 1741 | Intron | XP_016868653.1 | |||
XM_017013165.1 | 1741 | Intron | XP_016868654.1 |