Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGCCTCGCGCTCGAAGGCCGCC[C/T]TGTCCTCCAGGAAGCGCCGCTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BREA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BREA2 - breast cancer estrogen-induced apoptosis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC166 - coiled-coil domain containing 166 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162914.1 | 560 | Missense Mutation | AAG,AGG | K,R 187 | NP_001156386.1 |
LOC101928160 - uncharacterized LOC101928160 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK15 - mitogen-activated protein kinase 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |