Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGCTGAGGGCTTAAAGGAGGT[C/T]GGACCTCTTGGCCACCCTGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUSD3 - sushi domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287005.1 | Intron | NP_001273934.1 | ||||
NM_001287006.1 | Intron | NP_001273935.1 | ||||
NM_001287007.1 | Intron | NP_001273936.1 | ||||
NM_001287008.1 | Intron | NP_001273937.1 | ||||
NM_145006.3 | Intron | NP_659443.1 | ||||
XM_011518358.1 | Intron | XP_011516660.1 | ||||
XM_011518359.1 | Intron | XP_011516661.1 | ||||
XM_011518361.2 | Intron | XP_011516663.1 | ||||
XM_011518362.1 | Intron | XP_011516664.1 | ||||
XM_017014448.1 | Intron | XP_016869937.1 | ||||
XM_017014449.1 | Intron | XP_016869938.1 | ||||
XM_017014450.1 | Intron | XP_016869939.1 | ||||
XM_017014451.1 | Intron | XP_016869940.1 |