Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGATGCTGAGCCTTGACGCCT[C/T]GGCCATGGTCTACCCGCCGCGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRSAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005373.3 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | NP_001005373.1 | |
NM_001005374.3 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | NP_001005374.1 | |
NM_001190723.2 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | NP_001177652.1 | |
NM_138361.5 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | NP_612370.3 | |
XM_006717316.3 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | XP_006717379.1 | |
XM_017015283.1 | 1919 | Missense Mutation | TCG,TTG | S,L 183 | XP_016870772.1 | |
XM_017015284.1 | 1919 | UTR 5 | XP_016870773.1 |