Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTTTTTAGTGGTGAATTTCTC[A/G]GGGCAAATAAGGAGAAAGTGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610673 MIM: 608745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAIF1 - nuclear apoptosis inducing factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A25 - solute carrier family 25 member 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006641.3 | Intron | NP_001006642.1 | ||||
NM_001006642.3 | Intron | NP_001006643.1 | ||||
NM_001265614.2 | Intron | NP_001252543.1 | ||||
NM_052901.4 | Intron | NP_443133.2 | ||||
XM_005251688.1 | Intron | XP_005251745.1 | ||||
XM_005251689.4 | Intron | XP_005251746.1 | ||||
XM_006716948.2 | Intron | XP_006717011.1 | ||||
XM_006716949.3 | Intron | XP_006717012.1 |