Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATGGTCCAAAGGGACATGTCC[A/G]AGTCGCCTCCCACAGCGGCGGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600003 | ||||||||||||||||||||
Literature Links: |
CACNB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNB2 - calcium voltage-gated channel auxiliary subunit beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000724.3 | 82 | Intron | NP_000715.2 | |||
NM_001167945.1 | 82 | Intron | NP_001161417.1 | |||
NM_201570.2 | 82 | Intron | NP_963864.1 | |||
NM_201571.3 | 82 | Intron | NP_963865.2 | |||
NM_201572.3 | 82 | Intron | NP_963866.2 | |||
NM_201590.2 | 82 | Intron | NP_963884.2 | |||
NM_201593.2 | 82 | Missense Mutation | AAG,GAG | K,E 8 | NP_963887.2 | |
NM_201596.2 | 82 | Missense Mutation | AAG,GAG | K,E 8 | NP_963890.2 | |
NM_201597.2 | 82 | Missense Mutation | AAG,GAG | K,E 8 | NP_963891.1 | |
XM_005252588.3 | 82 | Intron | XP_005252645.1 | |||
XM_005252591.3 | 82 | Intron | XP_005252648.1 | |||
XM_006717502.3 | 82 | Intron | XP_006717565.1 | |||
XM_011519659.2 | 82 | Intron | XP_011517961.1 | |||
XM_011519660.2 | 82 | Intron | XP_011517962.1 | |||
XM_017016625.1 | 82 | Intron | XP_016872114.1 |