Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCAGCAGAAGAAACACCTAGG[A/G]AAATTACGAGTCCTGCAAAAGAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610856 | ||||||||||||||||||||
Literature Links: |
ANKRD30A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD30A - ankyrin repeat domain 30A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052997.2 | 1112 | Missense Mutation | AAA,GAA | K,E 304 | NP_443723.2 | |
XM_011519757.2 | 1112 | Missense Mutation | AAA,GAA | K,E 360 | XP_011518059.1 | |
XM_011519758.2 | 1112 | Intron | XP_011518060.1 | |||
XM_017016883.1 | 1112 | Intron | XP_016872372.1 |