Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAACATAGACATGGTGGCGGGTT[C/T]TGGTATCTTCTCTTGCTTTTTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600367 | ||||||||||||||||||||
Literature Links: |
CSTF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSTF3 - cleavage stimulation factor subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033505.1 | 1383 | Intron | NP_001028677.1 | |||
NM_001033506.1 | 1383 | Intron | NP_001028678.1 | |||
NM_001326.2 | 1383 | Missense Mutation | AAA,AGA | K,R 406 | NP_001317.1 |